missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloroaniline, 98%

Catalog No. AAA1392230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1392230 250 g
AAA1392218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1392230 Supplier Thermo Scientific Chemicals Supplier No. A1392230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 626-43-7 | C6H5Cl2N | 162.013 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 626-43-7
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007774
InChI Key UQRLKWGPEVNVHT-UHFFFAOYSA-N
Synonym benzenamine, 3,5-dichloro, m-dichloroaniline, aniline, 3,5-dichloro, 3,5-dichlorobenzenamine, 3,5-dichloro benzenamine, unii-oz75zm1s3g, 3,5-dichloranilin, 3,5 dichloraniline, ccris 2396, 3,5-dichloro aniline
PubChem CID 12281
ChEBI CHEBI:19904
IUPAC Name 3,5-dichloroaniline
SMILES C1=C(C=C(C=C1Cl)Cl)N

Specifications

Melting Point 49°C to 53°C
Density 1.58
Boiling Point 259°C to 260°C (741 mmHg)
Flash Point 133°C (271°F)
Quantity 250 g
UN Number UN3442
Beilstein 636492
Formula Weight 162.02
Percent Purity 98%
Chemical Name or Material 3,5-Dichloroaniline
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber 210-948-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only