missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-alpha-toluenesulfonyl chloride, 96%

Catalog No. AAH50707MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50707MD 250 mg
AAH5070703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50707MD Supplier Thermo Scientific Chemicals Supplier No. H50707MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 163295-70-3 | C7H5Cl3O2S | 259.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163295-70-3
Molecular Formula C7H5Cl3O2S
Molecular Weight (g/mol) 259.53
MDL Number MFCD01631928
InChI Key GZMJRPMBVICCFL-UHFFFAOYSA-N
Synonym 3,5-dichlorophenyl methanesulfonyl chloride, 3,5-dichloro-phenyl-methanesulfonyl chloride, acmc-1c80o, 3,5-dichloro-, a-toluenesufonyl chloride, 3,5-dichlorophenyl methyl chlorosulfone, 3,5-dichlorophenyl methanesulfonylchloride, 3,5-dichloro-alpha-toluenesulfonyl chloride, 3,5-dichlorophenyl methane-sulfonyl chloride, benzenemethanesulfonylchloride, 3,5-dichloro, 3,5-dichlorophenyl methyl sulfonyl chloride
PubChem CID 4993270
IUPAC Name (3,5-dichlorophenyl)methanesulfonyl chloride
SMILES ClC1=CC(CS(Cl)(=O)=O)=CC(Cl)=C1

Specifications

Quantity 250 mg
UN Number UN3096
Formula Weight 259.54
Percent Purity 96%
Chemical Name or Material 3,5-Dichloro-alpha-toluenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only