missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB03983DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. BTB03983DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

175135-06-5
417.034
YNPAAILIMIGQIH-UHFFFAOYSA-N
2774361
C1=CC(=C(C=C1[N+](=O)[O-])Cl)OC2=C(C=C(C=C2Cl)S(=O)(=O)Cl)Cl
C12H5Cl4NO5S
MFCD00052029
3,5-dichloro-4-2-chloro-4-nitrophenoxy benzene-1-sulfonyl chloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichloro-benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichlorobenzenesulfonyl chloride, benzenesulfonylchloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy
3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride

Specifications

3, 5-Dichloro-4-(2-chloro-4-nitrophenoxy)benzene-1-sulfonyl chloride
96.5
97%
C12H5Cl4NO5S
MFCD00052029
YNPAAILIMIGQIH-UHFFFAOYSA-N
3,5-dichloro-4-(2-chloro-4-nitrophenoxy)benzenesulfonyl chloride
2774361
97%
175135-06-5
100.0
Amber Glass Bottle
1g
3,5-dichloro-4-2-chloro-4-nitrophenoxy benzene-1-sulfonyl chloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichloro-benzenesulfonyl chloride, 4-2-chloro-4-nitrophenoxy-3,5-dichlorobenzenesulfonyl chloride, benzenesulfonylchloride, 3,5-dichloro-4-2-chloro-4-nitrophenoxy
C1=CC(=C(C=C1[N+](=O)[O-])Cl)OC2=C(C=C(C=C2Cl)S(=O)(=O)Cl)Cl
417.034
417.05
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents