missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromosalicylic acid, 97%, Thermo Scientific™

Catalog No. AC112920250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112920250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112920250 Supplier Thermo Scientific Chemicals Supplier No. 112920250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3147-55-5
Molecular Formula C7H4Br2O3
Molecular Weight (g/mol) 295.9
InChI Key BFBZHSOXKROMBG-UHFFFAOYSA-N
Synonym 3,5-dibromosalicylic acid, benzoic acid, 3,5-dibromo-2-hydroxy, 2-hydroxy-3,5-dibromobenzoic acid, unii-t5jo5ua21h, salicylic acid, 3,5-dibromo, t5jo5ua21h, rarechem al bo 2002, attercop-chm at113669, labotest-bb lt00455167, labotest-bb lt03333482
PubChem CID 18464
IUPAC Name 3,5-dibromo-2-hydroxybenzoic acid
SMILES C1=C(C=C(C(=C1Br)O)C(=O)O)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 225°C to 228°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 10,109
Merck Index 15,3033
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,ether
Formula Weight 295.9
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 5-Dibromosalicylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-570-9

RUO – Research Use Only