missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dibromoaniline 98.0+%, TCI America™

Catalog No. D48025G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D48025G Supplier TCI America Supplier No. D48025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 626-40-4
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.921
MDL Number MFCD00047841
InChI Key RVNUUWJGSOHMRR-UHFFFAOYSA-N
Synonym 3,5-dibromophenylamine, 3,5-dibromanilin, benzenamine, 3,5-dibromo, 1-amino-3,5-dibromobenzene, 3,5-dibromobenzeneamine, 3,5-dibromoamiline, 3,5-di-bromo-aniline, 3,5-dibromo-phenylamine, acmc-1b0ij, ksc493e8l
PubChem CID 221512
IUPAC Name 3,5-dibromoaniline
SMILES C1=C(C=C(C=C1Br)Br)N

Specifications

Melting Point 54°C
Quantity 5 g
Formula Weight 250.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dibromoaniline