missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylbenzoic acid, 99%

Catalog No. AC438220050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC438220050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC438220050 Supplier Thermo Scientific Chemicals Supplier No. 438220050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 16225-26-6 | C15H22O2 | 234.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16225-26-6
Molecular Formula C15H22O2
Molecular Weight (g/mol) 234.34
InChI Key NCTSLPBQVXUAHR-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylbenzoic acid, 3,5-di-t-butylbenzoic acid, 3,5-ditertbutylbenzoic acid, 3,5-di-tert-butylbenzoicacid, 3,5-bis tert-butyl benzoic acid, benzoic acid, 3,5-bis 1,1-dimethylethyl, pubchem4953, acmc-209do2, 3,5-di-tertbutylbenzoic acid, ksc494i2n
PubChem CID 85339
IUPAC Name 3,5-ditert-butylbenzoic acid
SMILES CC(C)(C)C1=CC(=CC(=C1)C(=O)O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 170.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Formula Weight 234.34
Percent Purity 99%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 3, 5-Di-tert-butylbenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 240-350-3

RUO – Research Use Only