missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 98%

Catalog No. AAB2041214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2041214 25 g
AAB2041206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2041214 Supplier Thermo Scientific Chemicals Supplier No. B2041214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20170-32-5 | C17H26O3 | 278.392 g/mol

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 20170-32-5
MDL Number MFCD00017519
Synonym 3-3,5-di-tert-butyl-4-hydroxyphenyl propionic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl propanoic acid, fenozan, 3,5-di-tert-butyl-4-hydroxyphenylpropionic acid, benzenepropanoic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxy, unii-x6g09g700f, 3,5-di-tert-butyl-4-hydroxyhydocinnamic acid, 3,5-bis 1,1-dimethylethyl-4-hydroxybenzenepropanoic acid, 3-3,5-ditertbutyl-4-hydroxyphenyl propionic acid, 3-3,5-bis tert-butyl-4-hydroxyphenyl propanoic acid
PubChem CID 88389
IUPAC Name 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoic acid

Specifications

Melting Point 172°C to 175°C
Molecular Formula C17H26O3
Quantity 25 g
Solubility Information Slightly soluble in water.
InChI Key WPMYUUITDBHVQZ-UHFFFAOYSA-N
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)CCC(=O)O
Molecular Weight (g/mol) 278.392
Formula Weight 278.4
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 243-556-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only