missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)styrene, 96%, stab.

Catalog No. AAB2349906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2349906 5 g
AAB2349903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2349906 Supplier Thermo Scientific Chemicals Supplier No. B2349906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 349-59-7 | C10H6F6 | 240.15 g/mol

3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Bis(trifluoromethyl)styrene is used to produce other chemicals for example 1-(3, 5-bis-Trifluoromethyl-phenyl)-ethane-1, 2-diol.

Solubility
Sparingly soluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 349-59-7
Molecular Formula C10H6F6
Molecular Weight (g/mol) 240.15
MDL Number MFCD00075538
InChI Key LFICVUCVPKKPFF-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl styrene, 1,3-bis trifluoromethyl-5-vinylbenzene, 1-ethenyl-3,5-bis trifluoromethyl benzene, 3,5-bis trifluoromethyl styrene, stab. with 4-methoxyphenol, pubchem2736, acmc-1cmtq, 3,5-bis trifluromethyl styrene, 1,3-bis-trifluoromethyl-5-vinyl-benzene, 3,5-bis trifluoromethyl styrene, stab.
PubChem CID 2773242
IUPAC Name 1-ethenyl-3,5-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(=CC(C=C)=C1)C(F)(F)F

Specifications

Melting Point 7°C to 8°C
Density 1.334
Boiling Point 59°C to 61°C (20 mmHg)
Flash Point 50°C (122°F)
Refractive Index 1.425
Quantity 5 g
UN Number UN1993
Solubility Information Sparingly soluble in water.
Formula Weight 240.15
Percent Purity 98%
Chemical Name or Material 3,5-Bis(trifluoromethyl)styrene, stabilized
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only