missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzoyl chloride, 97%

Catalog No. AC303530050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303530050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC303530050 Supplier Thermo Scientific Chemicals Supplier No. 303530050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 785-56-8 | C9H3ClF6O | 276.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 785-56-8
Molecular Formula C9H3ClF6O
Molecular Weight (g/mol) 276.57
MDL Number MFCD00000387
InChI Key WAKMMQSMEDJRRI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzoyl chloride, 3,5-bis-trifluoromethylbenzoyl chloride, 3,5-di trifluoromethyl benzoyl chloride, 3,5-bis trifluoromethyl benzoylchloride, benzoyl chloride, 3,5-bis trifluoromethyl, 3,5-bis-trifluoromethyl benzoyl chloride, benzoylchloride, 3,5-bis trifluoromethyl, fxffr cvg exfff, 3,5-bis trifluoromethyl benzoychloride
PubChem CID 101856
IUPAC Name 3,5-bis(trifluoromethyl)benzoyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5200g/mL
Boiling Point 65°C to 67°C (12.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4322 to 1.4342
Linear Formula (CF3)2C6H3COCl
Quantity 5 g
Specific Gravity 1.52
Formula Weight 276.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 5-Bis(trifluoromethyl)benzoyl chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-322-

RUO – Research Use Only