missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(4-aminophenoxy)benzoic Acid 96.0+%, TCI America™

Catalog No. B20815G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20815G 5 g
B20811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20815G Supplier TCI America Supplier No. B20815G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 195189-45-8
Molecular Formula C19H16N2O4
Molecular Weight (g/mol) 336.347
MDL Number MFCD02093443
InChI Key KPKOSOUTWDOOIW-UHFFFAOYSA-N
PubChem CID 11175081
IUPAC Name 3,5-bis(4-aminophenoxy)benzoic acid
SMILES C1=CC(=CC=C1N)OC2=CC(=CC(=C2)C(=O)O)OC3=CC=C(C=C3)N

Specifications

Melting Point 240°C
Color Yellow
Quantity 5 g
Formula Weight 336.35
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Bis(4-aminophenoxy)benzoic Acid