missing translation for 'onlineSavingsMsg'
Learn More

acide 3,5,7-Triméthyladamantane-1-carboxylique, 97%, Thermo Scientific™

Numéro de catalogue. BTB12973DA
Modifier l'affichage
Click to view available options
Quantité:
250 mg
1 g
Emballage:
Bouteille en verre ambrée
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
BTB12973DA 1 g Bouteille en verre ambrée
BTB12973CB 250 mg Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. BTB12973DA Fournisseur Thermo Scientific Chemicals Code fournisseur BTB12973DA

Identifiants chimiques

CAS 15291-66-4
Formule moléculaire C14H22O2
Poids moléculaire (g/mol) 222.33
Numéro MDL MFCD00187581
Clé InChI DHIIUPVOLLOORM-UHFFFAOYSA-N
Synonyme 3,5,8-trimethyladamantanecarboxylic acid, maybridge4_000576, 1,3,5-trimethyladamantane-7-carboxylic acid, 3,5,7-trimethyl adamantane-1-carboxylic acid, 1s,3r,5s,7s-3,5,7-trimethyladamantane-1-carboxylic acid, 3r,5s,7s-3,5,7-trimethyladamantane-1-carboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylicacid, 3,5,7-trimethyl, tricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, 3,5,7-trimethyl
PubChem CID 2802268
Nom de l’IUPAC 3,5,7-triméthyladamantane-1-acide carboxylique
SOURIRES CC12CC3(C)CC(C)(C1)CC(C2)(C3)C(O)=O

Spécifications

Nom ou substance chimique 3, 5, 7-Trimethyladamantane-1-carboxylic acid
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 1 g
Poids de formule 222.33
Pourcentage de pureté 97%
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.