missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethylhexanoic acid, 97%

Catalog No. AAL13360AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13360AE 100 mL
AAL13360AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL13360AE Supplier Thermo Scientific Chemicals Supplier No. L13360AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3302-10-1 | C9H18O2 | 158.24 g/mol

3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with the usual organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 3302-10-1
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00020507
InChI Key OILUAKBAMVLXGF-UHFFFAOYNA-N
Synonym hexanoic acid, 3,5,5-trimethyl, 3,5,5-trimethyl-hexanoic acid, 3,5,5-trimethylhexanoicacid, unii-aw943q219o, acmc-1assi, dsstox_cid_9254, 3,5-trimethylhexanoic acid, dsstox_rid_78735, dsstox_gsid_29254, 3,5,5 trimethylhexanoic acid
PubChem CID 90960
IUPAC Name 3,5,5-trimethylhexanoic acid
SMILES CC(CC(=O)O)CC(C)(C)C

Specifications

Melting Point -70°C
Density 0.91
Boiling Point 230°C to 232°C
Flash Point 125°C (257°F)
Odor Unpleasant
Refractive Index 1.429
Quantity 100 mL
Beilstein 1749506
Solubility Information Miscible with the usual organic solvents.
Formula Weight 158.24
Percent Purity 97%
Chemical Name or Material 3,5,5-Trimethylhexanoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-975-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only