missing translation for 'onlineSavingsMsg'
Learn More

3,5,5-Trimethylhexanal 90.0+%, TCI America™

Catalog No. p-7136509
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T094025ML 25 mL
T0940500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T094025ML Supplier TCI America Supplier No. T094025ML

Chemical Identifiers

CAS 5435-64-3
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00010441
InChI Key WTPYRCJDOZVZON-UHFFFAOYNA-N
Synonym 3,5,5-Trimethylhexanaldehyde
PubChem CID 21574
IUPAC Name 3,5,5-trimethylhexanal
SMILES CC(CC=O)CC(C)(C)C

Specifications

Color Yellow
Boiling Point 167°C
Quantity 25 mL
UN Number 1989
Formula Weight 142.24
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5,5-Trimethylhexanal
EINECSNumber (2)-0494
RTECSNumber MN7820000
TSCA Yes