missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)acrylic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 391750050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC391750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

5337-79-1
149.15
SSAYTINUCCRGDR-OWOJBTEDSA-N
736782
C1=CN=CC=C1C=CC(=O)O
C8H7NO2
MFCD00040746
3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
(E)-3-pyridin-4-ylprop-2-enoic acid

Specifications

3-(4-Pyridyl)acrylic acid
96.0
Authentic
Glass bottle
5g
3-4-pyridyl acrylic acid, trans-3-4-pyridyl acrylic acid, 3-pyridin-4-yl-acrylic acid, 4-pyridineacrylic acid, e-3-pyridin-4-yl acrylic acid, 3-pyridin-4-yl acrylic acid, 2e-3-pyridin-4-ylacrylic acid, 3-4-pyridine acrylic acid, 2e-3-pyridin-4-yl prop-2-enoic acid, e-3-pyridin-4-yl-acrylic acid
C1=CN=CC=C1C=CC(=O)O
149.15
149.15
5337-79-1
100.0
97%
C8H7NO2
MFCD00040746
SSAYTINUCCRGDR-OWOJBTEDSA-N
(E)-3-pyridin-4-ylprop-2-enoic acid
736782
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-265-4

SDS
Documents

Documents

Promotions

Promotions