missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Nitrophenyl)-1H-pyrazole, 97%, Thermo Scientific Chemicals

Catalog No. AAH3233204
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3233204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3233204 Supplier Thermo Scientific Chemicals Supplier No. H3233204
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20583-31-7 | Molecular Formula: C9H7N3O2 | Molecular Weight: 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20583-31-7
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
MDL Number MFCD00665859
InChI Key IPIYADCDDIUVPS-UHFFFAOYSA-N
Synonym 3-4-nitrophenyl-1h-pyrazole, 3-4-nitrophenyl pyrazole, 5-4-nitrophenyl-1h-pyrazole, 3-4-nitro-phenyl-1h-pyrazole, 5-4-nitrophenyl pyrazole, 1h-pyrazole, 3-4-nitrophenyl, 5-4-nitro-phenyl-1h-pyrazole, 3-4-nitrophenyl-2h-pyrazole, nitrophenylpyrazole, p-nitrophenylpyrazole
PubChem CID 2737071
IUPAC Name 5-(4-nitrophenyl)-1H-pyrazole
SMILES [O-][N+](=O)C1=CC=C(C=C1)C1=CC=NN1

Specifications

Melting Point 194°C to 195°C
Quantity 2 g
Formula Weight 189.18
Percent Purity 97%
Chemical Name or Material 3-(4-Nitrophenyl)-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only