missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Isopropylphenyl)isobutyraldehyde 92.0+%, TCI America™

Catalog No. I037725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I037725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I037725ML Supplier TCI America Supplier No. I037725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 103-95-7
Molecular Formula C13H18O
Molecular Weight (g/mol) 190.29
MDL Number MFCD00024160
InChI Key HQQPOVNESMNPNH-UHFFFAOYNA-N
Synonym 4-Isopropyl-alpha-methylhydrocinnamaldehyde
PubChem CID 517827
IUPAC Name 3-[4-(propan-2-yl)phenyl]butanal
SMILES CC(C)C1=CC=C(C=C1)C(C)CC=O

Specifications

Color Yellow
Boiling Point 270°C
Quantity 25 mL
Formula Weight 190.29
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(4-Isopropylphenyl)isobutyraldehyde
EINECSNumber (3)-2667
RTECSNumber MW4900000
TSCA Yes