missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionic acid, 99%

Catalog No. AAA1456730
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1456730 250 g
AAA1456709 10 g
AAA1456718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1456730 Supplier Thermo Scientific Chemicals Supplier No. A1456730
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 501-97-3 | C9H10O3 | 166.176 g/mol

3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(4-Hydroxyphenyl)propionic acid is a potential antioxidant. It is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 501-97-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002778
InChI Key NMHMNPHRMNGLLB-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid
PubChem CID 10394
ChEBI CHEBI:32980
IUPAC Name 3-(4-hydroxyphenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)O

Specifications

Melting Point 126°C to 131°C
Quantity 250 g
Beilstein 2209841
Solubility Information Soluble in water.
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material 3-(4-Hydroxyphenyl)propionic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-931-3
RTECSNumber MW5342000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only