missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dinitrobenzoic Acid 98.0+%, TCI America™

Catalog No. D08235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D08235G 5 g
D082325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D08235G Supplier TCI America Supplier No. D08235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 528-45-0
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007255
InChI Key OMVRRHJJQILIJX-UHFFFAOYSA-N
Synonym 3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956
PubChem CID 10709
IUPAC Name 3,4-dinitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 165°C
Color Yellow
Quantity 5 g
Formula Weight 212.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dinitrobenzoic Acid
EINECSNumber (3)-1505
RTECSNumber DG9140650
TSCA Yes