missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dinitrobenzoic acid, 99%

Catalog No. AC408700050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408700050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC408700050 Supplier Thermo Scientific Chemicals Supplier No. 408700050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 528-45-0 | C7H4N2O6 | 212.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 528-45-0
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007255
InChI Key OMVRRHJJQILIJX-UHFFFAOYSA-N
Synonym 3,4-dinitrobenzoic acid, benzoic acid, 3,4-dinitro, unii-0l3n4t0gpd, ccris 3128, 3,4-dinitrobenzioc acid, 0l3n4t0gpd, 3,4-dinitrobenzoate, 3,4-di-nitro-benzoic acid, zlchem 522, pubchem4956
PubChem CID 10709
IUPAC Name 3,4-dinitrobenzoic acid
SMILES OC(=O)C1=CC=C(C(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 163°C to 166°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (O2N)2C6H3CO2H
Quantity 5 g
Beilstein 09, 413
Fieser 01,320
Merck Index 15, 3301
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol,ether,hot water
Formula Weight 212.12
Percent Purity 99%
Chemical Name or Material 3, 4-Dinitrobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 208-433-9

RUO – Research Use Only