missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylphenylacetic acid, 98%

Catalog No. AAH6317806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6317806 5 g
AAH6317803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6317806 Supplier Thermo Scientific Chemicals Supplier No. H6317806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17283-16-8 | C10H11O2 | 163.20 g/mol

3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylphenylacetic acid, is is used as an intermediate in organic synthesis. It is also used as a fine chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 17283-16-8
Molecular Formula C10H11O2
Molecular Weight (g/mol) 163.20
MDL Number MFCD02664684
InChI Key OTTPBKINJOYJGW-UHFFFAOYSA-M
PubChem CID 296013
IUPAC Name 2-(3,4-dimethylphenyl)acetic acid
SMILES CC1=CC=C(CC([O-])=O)C=C1C

Specifications

Quantity 5 g
Physical Form Solid
Synonym 2-3,4-dimethylphenyl acetic acid, 3,4-dimethylphenylacetic acid, 3,4-dimethyl-phenyl-acetic acid, 3,4-dimethylphenylaceticacid, 3,4-dimethyphenylacetic acid, 3,4-dimethylphenyl acetic acid, benzeneacetic acid, 3,4-dimethyl, 3,4-dimethylbenzenacetic acid, pubchem10440, benzeneacetic acid,3,4-dimethyl
Solubility Information Slightly soluble in water.
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only