missing translation for 'onlineSavingsMsg'
Learn More

3,4-DimethylHexane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020247
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020247 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020247 Supplier Merck Emd Millipore Supplier No. 405121MLF
Petroleum

Specifications

CAS 583-48-2
Density 0.72 g/mL (at 25°C)
Flash Point 6°C
Physical Form Neat
Linear Formula CH3 CH2 CH(CH3)CH(CH3)CH2 CH3
MDL Number MFCD00039927
Boiling Point 119°C
Type Analytical Standard
Quantity 1 mL
Formula Weight 114.23
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less