missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzophenone 98.0+%, TCI America™

Catalog No. D328925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D328925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D328925G Supplier TCI America Supplier No. D328925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2571-39-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00008525
InChI Key JENOLWCGNVWTJN-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzophenone, 3,4-dimethylphenyl phenyl methanone, methanone, 3,4-dimethylphenyl phenyl, 3,4-dimethylphenyl-phenylmethanone, 3,4-dimethylphenyl phenyl ketone, acmc-1cbaf, 3,4-dimethyl-benzophenone, ksc490c7t, benzophenone, 3,4-dimethyl
PubChem CID 75730
IUPAC Name (3,4-dimethylphenyl)(phenyl)methanone
SMILES CC1=CC=C(C=C1C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 45°C
Color White-Yellow
Boiling Point 335°C
Quantity 25 g
Formula Weight 210.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethylbenzophenone