missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzophenone, 99%

Catalog No. AAB2096522
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2096522 100 g
AAB2096514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2096522 Supplier Thermo Scientific Chemicals Supplier No. B2096522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2571-39-3 | C15H14O | 210.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2571-39-3
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.28
MDL Number MFCD00008525
InChI Key JENOLWCGNVWTJN-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzophenone, 3,4-dimethylphenyl phenyl methanone, methanone, 3,4-dimethylphenyl phenyl, 3,4-dimethylphenyl-phenylmethanone, 3,4-dimethylphenyl phenyl ketone, acmc-1cbaf, 3,4-dimethyl-benzophenone, ksc490c7t, benzophenone, 3,4-dimethyl
PubChem CID 75730
IUPAC Name (3,4-dimethylphenyl)-phenylmethanone
SMILES CC1=CC=C(C=C1C)C(=O)C1=CC=CC=C1

Specifications

Melting Point 44°C to 47°C
Boiling Point 335°C
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 1952043
Formula Weight 210.28
Percent Purity 99%
Chemical Name or Material 3,4-Dimethylbenzophenone
EINECSNumber 219-916-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only