missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenyl Isothiocyanate 98.0+%, TCI America™

Catalog No. I054125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I054125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I054125G Supplier TCI America Supplier No. I054125G
Only null left

Chemical Identifiers

CAS 33904-04-0
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.236
MDL Number MFCD00041075
InChI Key LHPZZVZPOZPDDB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenyl isothiocyanate, 3,4-dimethoxyphenylisothiocyanate, 4-isothiocyanato-1,2-dimethoxy-benzene, isothiocyanic acid 3,4-dimethoxyphenyl ester, 3,4-dimethoxybenzenisothiocyanate, acmc-1aeuy, 3,4 dimethoxyphenylisothiocyanate, lhpzzvzpozpddb-uhfffaoysa, #, 3,4-dimethoxy phenyl isothiocyanate
PubChem CID 141856
IUPAC Name 4-isothiocyanato-1,2-dimethoxybenzene
SMILES COC1=C(C=C(C=C1)N=C=S)OC

Specifications

Melting Point 48°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 195.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxyphenyl Isothiocyanate