missing translation for 'onlineSavingsMsg'
Learn More

(3,4-Dimethoxyphenyl)acetonitrile, 99+%

Catalog No. AC155260250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC155260250 25 g Glass bottle
AC155261000 100 g Glass bottle
AC155265000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC155260250 Supplier Thermo Scientific Chemicals Supplier No. 155260250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 93-17-4 | C10H11NO2 | 177.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-17-4
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.2
MDL Number MFCD00001911
InChI Key ASLSUMISAQDOOB-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenylacetonitrile, 3,4-dimethoxyphenyl acetonitrile, homoveratronitrile, 3,4-dimethoxybenzyl cyanide, veratryl cyanide, 2-3,4-dimethoxyphenyl acetonitrile, 3,4-dimethoxybenzeneacetonitrile, benzeneacetonitrile, 3,4-dimethoxy, unii-62c8s706sp, 3,4-dimethoxy phenylacetonitrile
PubChem CID 66727
IUPAC Name 2-(3,4-dimethoxyphenyl)acetonitrile
SMILES COC1=C(C=C(C=C1)CC#N)OC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 62°C to 65°C
Color White
Boiling Point 171°C to 178°C (10.0 mmHg)
Flash Point 250°C
Loss on Drying 0.5% max. (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2CN
Quantity 25 g
Beilstein 10, I, 198
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,chloroform and,ethanol
Formula Weight 177.2
Percent Purity 99+%
Physical Form Crystals or Crystalline Needles
Chemical Name or Material (3, 4-Dimethoxyphenyl)acetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-225-1

RUO – Research Use Only