missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzyl alcohol, 97%

Catalog No. AAA1339614
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1339614 25 g
AAA1339622 100 g
AAA1339636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1339614 Supplier Thermo Scientific Chemicals Supplier No. A1339614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 93-03-8 | C9H12O3 | 168.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
3,4-Dimethoxybenzyl alcohol used as the fuel of the microbial fuel cell (MFC) to generate power. Veratrole alcohol is the raw material for the synthesis of cyclotriveratrylene which is used in host-guest chemistry.

Solubility
Fully miscible with water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 93-03-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004638
InChI Key OEGPRYNGFWGMMV-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
PubChem CID 7118
ChEBI CHEBI:62150
IUPAC Name (3,4-dimethoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)OC

Specifications

Melting Point ∼22°C
Density 1.176
Boiling Point 297°C to 298°C
Flash Point >110°C (230°F)
Refractive Index 1.553
Quantity 25 g
Beilstein 639388
Solubility Information Fully miscible with water.
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 3,4-Dimethoxybenzyl alcohol
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 202-212-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only