missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. D0631500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0631500G 500 g
D063125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0631500G Supplier TCI America Supplier No. D0631500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-07-2
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.175
MDL Number MFCD00002500
InChI Key DAUAQNGYDSHRET-UHFFFAOYSA-N
Synonym veratric acid, benzoic acid, 3,4-dimethoxy, dimethylprotocatechuic acid, veratrylic acid, 3,4-dimethylprotocatechuic acid, veratrumenoic acid, 3,4-dimethoxy-benzoic acid, unii-1yy04e7rr4, 3,4-dimethoxybenzoes
PubChem CID 7121
ChEBI CHEBI:296881
IUPAC Name 3,4-dimethoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)OC

Specifications

Melting Point 182°C
Quantity 500 g
Formula Weight 182.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxybenzoic Acid
EINECSNumber (3)-1645
RTECSNumber DG8598750
TSCA Yes