missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyaniline 98.0+%, TCI America™

Catalog No. D148525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D148525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D148525G Supplier TCI America Supplier No. D148525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6315-89-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00008394
InChI Key LGDHZCLREKIGKJ-UHFFFAOYSA-N
Synonym 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
PubChem CID 22770
IUPAC Name 3,4-dimethoxyaniline
SMILES COC1=C(C=C(C=C1)N)OC

Specifications

Melting Point 87°C
Boiling Point 180°C
Quantity 25 g
Formula Weight 153.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxyaniline
EINECSNumber (3)-0616
RTECSNumber BX4550000
TSCA Yes