Learn More
3,4-Dimethoxyaniline, 98+%
CAS: 6315-89-5 | C8H11NO2 | 153.181 g/mol
Supplier: Thermo Scientific Chemicals A1354818
Description
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.
Solubility
Slightly soluble in water
Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
| 3,4-Dimethoxyaniline | |
| 86°C to 89°C | |
| >110°C (230°F) | |
| C8H11NO2 | |
| 50 g | |
| 743399 | |
| Slightly soluble in water | |
| COC1=C(C=C(C=C1)N)OC | |
| 153.181 | |
| 153.18 |
| 6315-89-5 | |
| 174°C to 176°C (22 mmHg) | |
| Characteristic | |
| MFCD00008394 | |
| UN2811 | |
| 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine | |
| LGDHZCLREKIGKJ-UHFFFAOYSA-N | |
| 3,4-dimethoxyaniline | |
| 22770 | |
| ≥98% |
Chemical Identifiers
| 6315-89-5 | |
| 153.181 | |
| LGDHZCLREKIGKJ-UHFFFAOYSA-N | |
| 22770 | |
| COC1=C(C=C(C=C1)N)OC |
| C8H11NO2 | |
| MFCD00008394 | |
| 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine | |
| 3,4-dimethoxyaniline |
Safety and Handling
GHS H Statement
H311-H373-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 228-647-6
RTECSNumber : BX4550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only