missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyaniline, 98+%

Catalog No. p-7021275
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1354809 10 g
AAA1354818 50 g
AAA1354830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1354809 Supplier Thermo Scientific Chemicals Supplier No. A1354809

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6315-89-5 | C8H11NO2 | 153.181 g/mol

3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyaniline is commonly used as fine chemicals, used for pharmaceutical intermediates.

Solubility
Slightly soluble in water

Notes
Air Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 6315-89-5
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00008394
InChI Key LGDHZCLREKIGKJ-UHFFFAOYSA-N
Synonym 4-aminoveratrole, benzenamine, 3,4-dimethoxy, 4-aminoveratrol, 3,4-dimethoxybenzenamine, 2-methoxy-4-aminoanisole, 3,4-dimethoxy aniline, aniline, 3,4-dimethoxy, 3,4-dimethyloxy aniline, 3,4-dimethoxy-phenylamine
PubChem CID 22770
IUPAC Name 3,4-dimethoxyaniline
SMILES COC1=C(C=C(C=C1)N)OC

Specifications

Melting Point 86°C to 89°C
Boiling Point 174°C to 176°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 10 g
UN Number UN2811
Beilstein 743399
Solubility Information Slightly soluble in water
Formula Weight 153.18
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxyaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H311-H373-H302-H332
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 228-647-6
RTECSNumber BX4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only