accessibility menu, dialog, popup

UserName

H3CONH2OCH3

CAS RN 6315-89-5

IUPAC Name: 3,4-dimethoxyaniline
Synonyms: benzenamine, 3,4-dimethoxy-
Molecular Weight (g/mol): 153.18
Molecular Formula: C8H11NO2
InChi Key: LGDHZCLREKIGKJ-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C=C1OC
Boiling Point 
  • (3)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)