missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethoxyacetophenone, 98%

Catalog No. AC168850250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168850250 25 g Glass bottle
AC168851000 100 g Glass bottle
AC168855000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC168850250 Supplier Thermo Scientific Chemicals Supplier No. 168850250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1131-62-0 | C10H12O3 | 180.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1131-62-0
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.2
MDL Number MFCD00008737
InChI Key IQZLUWLMQNGTIW-UHFFFAOYSA-N
Synonym 3',4'-dimethoxyacetophenone, 1-3,4-dimethoxyphenyl ethanone, 3,4-dimethoxyacetophenone, acetoveratrone, ethanone, 1-3,4-dimethoxyphenyl, 1-3,4-dimethoxyphenyl ethan-1-one, 3,4-dimethoxyphenyl methyl ketone, unii-5rv6436s8a, acetophenone, 3',4'-dimethoxy, acetophenone,4'-dimethoxy
PubChem CID 14328
ChEBI CHEBI:86576
IUPAC Name 1-(3,4-dimethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48.0°C to 52.0°C
Color Yellow to Beige
Boiling Point 286.0°C to 288.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3COCH3
Quantity 25 g
Beilstein 08, II, 298
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol
Formula Weight 180.2
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3', 4'-Dimethoxyacetophenone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautious
EINECSNumber 214-468-

RUO – Research Use Only