missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzonitrile 98.0+%, TCI America™

Catalog No. D33525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33525G 5 g
D335225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33525G Supplier TCI America Supplier No. D33525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17345-61-8
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.122
MDL Number MFCD00016436
InChI Key NUWHYWYSMAPBHK-UHFFFAOYSA-N
Synonym protocatachuonitrile, 3,4-dihydroxy benzonitrile, 3,4-dihydroxybenzenecarbonitrile, protocatechuonitrile, 4-cyanocatechol, protocatechunitrile, 4-cyanopyrocatechol, pubchem3120, acmc-209e6q, 3, 4-dihydroxybenzonitrile
PubChem CID 87065
IUPAC Name 3,4-dihydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)O)O

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 135.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dihydroxybenzonitrile