missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydroxybenzoic acid, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 99-50-3 |
| Linear Formula | (HO)2C6H3 CO2 H |
| MDL Number | MFCD00002509 |
| Melting Point | 197°C to 200°C |
| Synonym | Protocatechuic acid |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Formula Weight | 154.12 |
| Percent Purity | ≥97.0% (HPLC) |
| Show More |