missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydrocoumarin 98,0+%, TCI America™
Spécifications
Spécifications
| Color | Blanc |
| Boiling Point | 149°C |
| Quantity | 25 g |
| Formula Weight | 148.16 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Morceaux cristallins |
| Chemical Name or Material | 3,4-Dihydrocoumarin |
Sécurité et manipulation
| EINECSNumber | (5)-0689 |
| RTECSNumber | MW5775000 |
| TSCA | Yes |