missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-pyran, 99%

Numéro de catalogue. p-112285
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC114781000 100 g Glass bottle
AC114785000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC114781000 Fournisseur Thermo Scientific Chemicals Code fournisseur 114781000
Il en reste null

CAS: 110-87-2 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 110-87-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00006558
InChI Key BUDQDWGNQVEFAC-UHFFFAOYSA-N
Synonym dihydropyran, 3,4-dihydropyran, 2,3-dihydropyran, 2h-3,4-dihydropyran, 2,3-dihydro-4h-pyran, dihydro-2h-pyran, 2h-pyran, dihydro, 2h-pyran, 3,4-dihydro, 5,6-dihydro-4h-pyran, dihydropyrane
PubChem CID 8080
IUPAC Name 3,4-dihydro-2H-pyran
SMILES C1CC=COC1

Spécifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 87.0°C
Flash Point −9°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4405 to 1.4425
Quantity 100 g
UN Number 2376
Fieser 01,256; 03,99; 05,220; 07,109; 10,147; 11,26
Specific Gravity 0.92
Merck Index 15, 3195
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in ethanol,slightly soluble in chloroform,soluble in most other organic solvents
Viscosity 0.76 mPa.s (20°C)
Formula Weight 84.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 4-Dihydro-2H-pyran
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
EINECSNumber 203-810-4
TSCA TSCA

RUO – Research Use Only