missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-amine, 98%

Catalog No. AAH2786206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2786206 5 g
AAH2786203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2786206 Supplier Thermo Scientific Chemicals Supplier No. H2786206

CAS: 175136-34-2 | C9H11NO2 | 165.192 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water. High soluble in ethanol, acetone.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 175136-34-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00218806
InChI Key FVLCICVRAPEYNX-UHFFFAOYSA-N
PubChem CID 2775654
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-amine
SMILES C1COC2=C(C=C(C=C2)N)OC1

Specifications

Melting Point 79°C to 81°C
Boiling Point 123°C to 128°C (0.2 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine
Solubility Information Soluble in water. High soluble in ethanol,acetone.
Formula Weight 165.19
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin-7-amine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only