missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin-7-amine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 175136-34-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
MDL Number MFCD00218806
InChI Key FVLCICVRAPEYNX-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine
PubChem CID 2775654
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepin-7-amine
SMILES C1COC2=C(C=C(C=C2)N)OC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
BTB12798DA
View Documents Promotion Details
N/A 1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB12798EA
View Documents Promotion Details
N/A 10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

175136-34-2
165.192
FVLCICVRAPEYNX-UHFFFAOYSA-N
2775654
C1COC2=C(C=C(C=C2)N)OC1
C9H11NO2
MFCD00218806
3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine
3,4-dihydro-2H-1,5-benzodioxepin-7-amine

Specifications

175136-34-2
100.0
C9H11NO2
3,4-dihydro-2h-benzo b 1,4 dioxepin-7-amine, 2h-1,5-benzodioxepin-7-amine,3,4-dihydro, 2h-1,5-benzodioxepin-7-amine, 3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepin-7-ylamine, maybridge1_002016, acmc-1c5sw, buttpark 12115-34, 3,4-trimethylenedioxyaniline, 7-amino-3,4-dihydro-2h-1,5-benzodioxepine, 3, 4-dihydro-7-amino-2h-benzo b 1,4 dioxepine
C1COC2=C(C=C(C=C2)N)OC1
165.192
165.19
3, 4-Dihydro-2H-1, 5-benzodioxepin-7-amine
96.5
97%
MFCD00218806
FVLCICVRAPEYNX-UHFFFAOYSA-N
3,4-dihydro-2H-1,5-benzodioxepin-7-amine
2775654
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents