missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzylamine 98.0+%, TCI America™

Catalog No. p-7127698
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33731G 1 g
D33735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D33731G Supplier TCI America Supplier No. D33731G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72235-53-1
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.137
MDL Number MFCD00010145
InChI Key PHLZUDXEBCQHKM-UHFFFAOYSA-N
Synonym 3,4-difluorobenzylamine, 3,4-difluorophenyl methanamine, 3,4-difluorobenzyl amine, benzenemethanamine, 3,4-difluoro, 1-3,4-difluorophenyl methanamine, 3,4-difluorophenyl methylamine, 3, 4-difluorobenzylamine, pubchem1918, acmc-209omb, 3,4-difluoro-benzylamine
PubChem CID 123572
IUPAC Name (3,4-difluorophenyl)methanamine
SMILES C1=CC(=C(C=C1CN)F)F

Specifications

Quantity 1 g
UN Number 3267
Formula Weight 143.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,4-Difluorobenzylamine