missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzonitrile 98.0+%, TCI America™

Catalog No. D23415G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23415G 5 g
D234125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23415G Supplier TCI America Supplier No. D23415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64248-62-0
Molecular Formula C7H3F2N
Molecular Weight (g/mol) 139.11
MDL Number MFCD00011666
InChI Key BTBFCBQZFMQBNT-UHFFFAOYSA-N
Synonym benzonitrile, 3,4-difluoro, 4-cyano-1,2-difluorobenzene, 3,4-difluoro benzonitrile, 3,4-difluorobenzenecarbonitrile, pubchem1562, acmc-209nkz, 3,4 difluorobenzonitrile, 3,4-difluoro-benzonitrile, ksc355i7l, 4-fluoro-3-fluorobenzonitrile
PubChem CID 587203
IUPAC Name 3,4-difluorobenzonitrile
SMILES FC1=CC=C(C=C1F)C#N

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 95°C
Quantity 5 g
UN Number 3439
Formula Weight 139.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Difluorobenzonitrile
EINECSNumber (3)-4305
TSCA No