missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorobenzoic acid, 98%

Catalog No. AAA1587614
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1587614 25 g
AAA1587603 1 g
AAA1587606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1587614 Supplier Thermo Scientific Chemicals Supplier No. A1587614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-86-7 | C7H4F2O2 | 158.104 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 455-86-7
Molecular Formula C7H4F2O2
Molecular Weight (g/mol) 158.104
MDL Number MFCD00011672
InChI Key FPENCTDAQQQKNY-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-difluoro, 3,4-fifluorobenzoic acid, 3,4-difluoro-benzoic acid, 3,4-dichloropnenol, pubchem1337, acmc-209k3h, 3,4-difluoro benzoic acid, ksc237g8t, rarechem al bo 0269, timtec-bb sbb006721
PubChem CID 99166
IUPAC Name 3,4-difluorobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)F)F

Specifications

Melting Point 121°C to 124°C
Odor Odorless
Quantity 25 g
Beilstein 2085848
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material 3,4-Difluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-249-6
TSCA Yes

RUO – Research Use Only