missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenylacetic Acid 98.0+%, TCI America™

Catalog No. D1813500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1813500G 500 g
D181325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1813500G Supplier TCI America Supplier No. D1813500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5807-30-7
Molecular Formula C8H6Cl2O2
Molecular Weight (g/mol) 205.034
MDL Number MFCD00004333
InChI Key ZOUPGSMSNQLUNW-UHFFFAOYSA-N
Synonym 3,4-dichlorophenylacetic acid, 2-3,4-dichlorophenyl acetic acid, 3,4-dichlorophenyl acetic acid, 3,4-dichlorobenzene acetic acid, benzeneacetic acid, 3,4-dichloro, pubchem20195, acmc-209m2g, 3,4dichlorophenylacetic acid, 3,4-dichlorophenylaceticacid
PubChem CID 79874
IUPAC Name 2-(3,4-dichlorophenyl)acetic acid
SMILES C1=CC(=C(C=C1CC(=O)O)Cl)Cl

Specifications

Melting Point 90°C
Color White
Quantity 500 g
Formula Weight 205.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorophenylacetic Acid
EINECSNumber (3)-3805
TSCA No