missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenyl isocyanate, 97%

Catalog No. AAA1629806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1629806 5 g
AAA1629814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1629806 Supplier Thermo Scientific Chemicals Supplier No. A1629806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 102-36-3 | C7H3Cl2NO | 188.007 g/mol

Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C 'labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-labeled diuron was prepared by the reaction of 3,4-dichlorophenyl- isocyanate with C ′″labeled dimethylamine.Substituted urea derivatives were prepared by reacting 3,4-dichlorophenyl isocyanate with amino acids, dipeptides, histamine. The R- and S-isomers and RS-racemate of MBPU were synthesized by allowing equimolar quantities of 3,4-dichlorophenyl- isocyanate

Solubility
Hydrolyzes in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water, amines, alcohols.

Chemical Identifiers

CAS 102-36-3
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.007
MDL Number MFCD00002017
InChI Key MFUVCHZWGSJKEQ-UHFFFAOYSA-N
Synonym 3,4-dichlorophenyl isocyanate, benzene, 1,2-dichloro-4-isocyanato, 3,4-dichlorfenylisokyanat, isocyanic acid 3,4-dichlorophenyl ester, 3,4-dichlorophenylisocyanate, unii-ozw0b7f1du, 3,4-dichlorphenylisocyanate, isocyanic acid, 3,4-dichlorophenyl ester, 3,4-dichlorfenylisokyanat czech, ozw0b7f1du
PubChem CID 7607
IUPAC Name 1,2-dichloro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)Cl)Cl

Specifications

Melting Point 40°C to 44°C
Boiling Point 118°C to 120°C (18 mmHg)
Flash Point 123°C (253°F)
Odor Irritating
Quantity 5 g
UN Number UN2250
Beilstein 608325
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorophenyl isocyanate
Show More Show Less
Hazard Category H301-H312+H332-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H300-H331-H334-H315-H319-H335
Fatal if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROPHENYL ISOCYANATES
EINECSNumber 203-026-2
RTECSNumber NQ8760000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only