missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorophenol, 97%

Catalog No. AAA1225622
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1225622 100 g
AAA1225614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1225622 Supplier Thermo Scientific Chemicals Supplier No. A1225622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 95-77-2 | C6H4Cl2O | 163.00 g/mol

It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is also a photo-degradation product of the common antibacterial and antifungal agent triclosan along with the dioxin 2,8-dichlorodibenzo-p-dioxin.

Solubility
Slightly soluble ion water 9.2 g/L at 20degC. Soluble in aqueous alkali, oxygenated and chlorinated solvents. Soluble in ethanol, benzene, chloroform and ethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents, acid, acid fumes, acid chlorides, acid anhydrides, caustics. Keep container tightly closed. Store in an area without drain or sewer access. Separated from strong oxidants and food and feedstuffs.

Chemical Identifiers

CAS 95-77-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002258
InChI Key WDNBURPWRNALGP-UHFFFAOYSA-N
PubChem CID 7258
ChEBI CHEBI:34323
IUPAC Name 3,4-dichlorophenol
SMILES OC1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 64°C to 68°C
Boiling Point 252°C to 255°C
Odor Phenol-like
Quantity 100 g
UN Number UN2020
Beilstein 1907693
Synonym 4,5-dichlorophenol, phenol, 3,4-dichloro, 3,3-dichloro-hydroxybenzene, unii-070ftm6dvf, ccris 5904, 070ftm6dvf, 3,4-dcp, 3,4-dichlorphenol, 3,4dichlorophenol, 3,4 dichlorophenol
Solubility Information Slightly soluble ion water 9.2g/L at 20degC. Soluble in aqueous alkali,oxygenated and chlorinated solvents. Soluble in ethanol,benzene,chloroform and ethyl ether.
Formula Weight 163
Percent Purity 99%
Chemical Name or Material 3,4-Dichlorophenol
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROPHENOLS, SOLID
EINECSNumber 202-450-5
RTECSNumber SK8800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only