missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dichlorobenzotrichloride 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 500 g |
| UN Number | 1759 |
| Formula Weight | 264.35 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 3,4-Dichlorobenzotrichloride |
Safety and Handling
| EINECSNumber | (3)-0093 |
| RTECSNumber | CZ5475000 |
| TSCA | Yes |