missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzonitrile 98.0+%, TCI America™

Catalog No. D34155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34155G 5 g
D341525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34155G Supplier TCI America Supplier No. D34155G
Only null left

Chemical Identifiers

CAS 6574-99-8
Molecular Formula C7H3Cl2N
Molecular Weight (g/mol) 172.008
MDL Number MFCD00016379
InChI Key KUWBYWUSERRVQP-UHFFFAOYSA-N
Synonym benzonitrile, 3,4-dichloro, 3,4-dichloro benzonitrile, 3.4-dichlorobenzonitrile, 3, 4-dichlorobenzonitrile, 3,4-dichlorobenzenecarbonitrile, 3,4-dichloro-benzonitrile, pubchem3688, acmc-1b9ly, 3,4-dichlorobenzonitrile, buttpark 87\02-72
PubChem CID 81051
IUPAC Name 3,4-dichlorobenzonitrile
SMILES C1=CC(=C(C=C1C#N)Cl)Cl

Specifications

Melting Point 73°C
Color White
Quantity 5 g
UN Number 3439
Formula Weight 172.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzonitrile