missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzamide 98.0+%, TCI America™

Catalog No. D44745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44745G 5 g
D44741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44745G Supplier TCI America Supplier No. D44745G
Only null left

Chemical Identifiers

CAS 2670-38-4
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.02
MDL Number MFCD00053009
InChI Key XURBWYCGQQXTHJ-UHFFFAOYSA-N
Synonym benzamide, 3,4-dichloro, acmc-20anfl, pubchem3462, 3,4-dichloro benzamide, benzamide,3,4-dichloro, 3,4-bis chloranyl benzamide
PubChem CID 137671
IUPAC Name 3,4-dichlorobenzamide
SMILES NC(=O)C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 144°C
Color White
Quantity 5 g
Formula Weight 190.02
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dichlorobenzamide