missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibromothieno[2,3-b]thiophene 98.0+%, TCI America™

Catalog No. D40331G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D40331G Supplier TCI America Supplier No. D40331G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 53255-78-0
Molecular Formula C6H2Br2S2
Molecular Weight (g/mol) 298.01
InChI Key BUGQRBSPOFSLQS-UHFFFAOYSA-N
PubChem CID 143030
IUPAC Name 3,4-dibromothieno[2,3-b]thiophene
SMILES C1=C(C2=C(S1)SC=C2Br)Br

Specifications

Melting Point 133°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dibromothieno[2,3-b]thiophene
TSCA No
Recommended Storage Freezer