missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-1-propanol, 98%

Catalog No. AAH6111806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6111806 5 g
AAH6111803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6111806 Supplier Thermo Scientific Chemicals Supplier No. H6111806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 25574-11-2 | C9H11BrO | 215.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25574-11-2
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD09028724
InChI Key WODKXGCVVOOEIJ-UHFFFAOYSA-N
Synonym 3-4-bromophenyl propan-1-ol, 4-bromobenzenepropanol, 3-4-bromo-phenyl-propan-1-ol, benzenepropanol, 4-bromo, 3-4-bromophenyl-1-propanol, pubchem3984, 3-4-bromophenyl propanol, 3-4-bromophenyl-propanol, amth183, 3-4-bromophenyl propyl alcohol
PubChem CID 10560614
IUPAC Name 3-(4-bromophenyl)propan-1-ol
SMILES C1=CC(=CC=C1CCCO)Br

Specifications

Linear Formula BrC6H4(CH2)3OH
Quantity 5 g
Formula Weight 215.09
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-(4-Bromophenyl)-1-propanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only