missing translation for 'onlineSavingsMsg'
Learn More

3,4,9,10-Perylenetetracarboxylic Dianhydride (purified by sublimation) 98.0+%, TCI America™

Catalog No. P21021G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P21021G Supplier TCI America Supplier No. P21021G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 128-69-8
Molecular Formula C24H8O6
Molecular Weight (g/mol) 392.32
MDL Number MFCD00006916
InChI Key CLYVDMAATCIVBF-UHFFFAOYSA-N
Synonym 3,4,9,10-perylenetetracarboxylic dianhydride, pigment red 224, ptcda, perylene-3,4,9,10-tetracarboxylic dianhydride, perylenetetracarboxylic anhydride, anthra 2,1,9-def:6,5,10-d'e'f' diisochromene-1,3,8,10-tetraone, perylo 3,4-cd:9,10-c'd' dipyran-1,3,8,10-tetrone, perylenetetracarboxylic acid dianhydride, 3,4:9,10-perylenetetracarboxylic anhydride
PubChem CID 67191
IUPAC Name 7,18-dioxaheptacyclo[14.6.2.2²,⁵.0³,¹².0⁴,⁹.0¹³,²³.0²⁰,²⁴]hexacosa-1(22),2(26),3,5(25),9,11,13,15,20,23-decaene-6,8,17,19-tetrone
SMILES O=C1OC(=O)C2=CC=C3C4=CC=C5C(=O)OC(=O)C6=CC=C(C7=CC=C1C2=C37)C4=C56

Specifications

Quantity 1 g
Formula Weight 392.32
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,9,10-Perylenetetracarboxylic Dianhydride (purified by sublimation)
EINECSNumber (4)-1307&(9)-2398
TSCA Yes